Learn More
Brensocatib, MedChemExpress
Product Code. 16463248
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463248 10 mg 10mg
16482686 25 mg 25mg
16439796 50 mg 50mg
16434516 100 mg 100mg
16425977 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16463248 Supplier Medchem Express Supplier No. HY101056
MedChemExpress Brensocatib (AZD7986) is an oral dipeptidyl peptidase 1 (DPP1) inhibitor with pIC50s of 6.85, 7.6, 7.7, 7.8, and 7.8 in human, mouse, rat, dog and rabbit, respectively.
Brensocatib (AZD7986) is an oral dipeptidyl peptidase 1 (DPP1) inhibitor with pIC50s of 6.85, 7.6, 7.7, 7.8, and 7.8 in human, mouse, rat, dog and rabbit, respectively.
Specifications
Chemical Name or Material Brensocatib
CAS 1802148-05-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H24N4O4
Quantity 10 mg
Synonym AZD7986 INS 1007
Solubility Information DMSO : ≥ 100 mg/mL (237.83 mM)
SMILES O=C([C@H]1OCCCNC1)N[C@H](C#N)CC2=CC=C(C3=CC=C(OC(N4C)=O)C4=C3)C=C2
Molecular Weight (g/mol) 420.46
Formula Weight 420.46
Percent Purity 99.66%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation