Learn More
Fisherbrand™ Meker Steel Bunsen Burner Gas Head
Product Code. 11759317 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Material:
Steel
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
11759317 Steel Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11759317
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
Gas burner head only
Replaceable steel perforated head fits on brass base
Specifications
Product Type Gas Head
For Use With (Equipment) Meker Burner
Material Steel
No. per Pack 1 Ea.