Learn More
Branson Ultrasonics™ Sonifier™ Replacement Flat Tip
Product Code. 10337832
Change view
Click to view available options
For Use With (Equipment):
Sonifier™ cell disruptor
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10337832 Sonifier™ cell disruptor Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10337832 Supplier Branson Ultrasonics™ Supplier No. 101148013
  • ONLINE EXCLUSIVE PRICE
    119.00 EUR
    84.00 EUR / Pack of 1
    Save 35.00 EUR
    29% Off
Estimated Shipment: 09-12-2026
Tip for 1/2 in. probe.
Tip to use as a 1/2 in. probe, without tip probe used to attach tapered microtips.
Specifications
For Use With (Equipment) Sonifier™ cell disruptor
Tip Style Flat
Product Type Flat Microtip