Learn More
VACUUBRAND™ VSP3000 External Vacuum Sensor
Click to view available options
:
VSP3000 vacuum sensor
Description
- Based on thermal conductivity (Pirani) and offers a wide measurement range from atmospheric pressure down to 10-3 mbar
- Made of plastics and ceramics with high chemical resistance, splash-water proof, also for rough operating conditions
- Designed for use with VACUUBRAND DCP3000 Pirani fine vacuum gauge Fisher Cat 13-878-320
Specifications
Specifications
| For Use With (Equipment) | Vacuubrand Vacuubus VSP3000 Pirani Fine Vacuum Gauge |
| Product Type | Vacuum Sensor |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu