Learn More
Cole Parmer BrandTech Pipette Controller
Product Code. 15715072 Shop All Cole Parmer Products
Change view
Click to view available options
Volume (Metric):
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15715072 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15715072 Supplier Cole Parmer Supplier No. 2455049
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
BrandTech Pipette Controller is color-coded for quick identification
  • Measure and dispense with a single lever
  • Blow-out button clears the pipette
  • Adapter fits all pipettes from 0.1 to 100 mL
  • Membrane filter protects controller from fumes and liquids
  • Filter and adapter may be autoclaved
Specifications
Color Natural
Product Line BrandTech
Sterility Non-sterile
Type Pipette Controller
Volume (Metric) 100 mL