Description
- Maintains sample temperature at 4°C for approximately 3 hours
- Insulating gel changes from violet to pink at 7°C for easy, visual monitoring
- Compatible with 0.2 mL tubes, 8-tube strips, 12-tube strips, and 96-well PCR plates
- Ideal for preventing sample warming during setup and handling
- Transparent lid included
- Sold in packs of 2
Specifications
Specifications
| Dimensions (L x W x H) | 140 x 98 x 45 mm |
| Includes | Transparent lid |
| Material | Polypropylene |
| Product Type | Mini Cooler |
| Color | Pink |
| Capacity (Metric) | 19.2 mL |
| Autoclavable | Not Autoclavable |
| Temperature Control | Passive |
| Temperature Range | 2°C to 8°C |