Learn More
BRAND™ Sealing paddle for sealing films
Product Code. 15708987 Shop All Brand Products
Change view
Click to view available options
Quantity:
2/Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15708987 2/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15708987 Supplier BRAND™ Supplier No. 701381
  • ONLINE EXCLUSIVE PRICE
    6.75 EUR
    5.66 EUR / Pack of 1
    Save 1.09 EUR
    16% Off
Estimated Shipment: 21-12-2026
for affixing plate sealing films
  • Assist in effective adhering of sealing films on microplates, PCR plates and deepwell plates
  • Package of 2
TRUSTED_SUSTAINABILITY
Specifications
Description Sealing paddle for affixing plate sealing films
For Use With (Equipment) Microplates, deep-well plates
No. per Pack 2/Pk.
Product Type Sealing Paddle
Quantity 2/Pk.