Learn More
Brand™ Natural Rubber Cap
Product Code. 12090912 Shop All Brand Products
Change view
Click to view available options
Quantity:
100/Pk.
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12090912 100/Pk. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12090912 Supplier Brand™ Supplier No. 124700
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
Bulk Quote Request
Assists with liquid/media retrieval and pipetting of Pasteur pipets and dropping bottles. BRAND™ Natural Rubber Cap fits a variety of pipets and provides more accurate dropping.
Pipetting aid for glass Pasteur pipets and dropping bottles
TRUSTED_SUSTAINABILITY
Specifications
Material Natural Rubber
For Use With (Application) Gass Pasteur pipets and dropping bottles
Quantity 100/Pk.
Type Cap
For Use With (Equipment) Pasteur Pipet