Learn More
Brady™ B30 Series R6400 Printer Ribbon, Black
Product Code. 13503310 Shop All Brady Products
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
13503310 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13503310 Supplier Brady™ Supplier No. 143118
  • ONLINE EXCLUSIVE PRICE
    94.25 EUR
    87.96 EUR / Pack of 1
    Save 6.29 EUR
    6% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Tough, smear-proof, drop and clink print ribbons are used for quick and easy supply changes.
B30 Series Drop & Click print ribbons enable 5-second ribbon supply changes. No calibrating, no threading, no waste, no hassles and the ink always faces the right direction!
  • One-color ribbon for printing a single color onto a label
  • R6400 Series Resin Formulation offers superior chemical and abrasion resistance when printed on the appropriate label material from Brady™
  • Recommended for use with materials designed to resist chemicals, such as B-492 Frozen Surface Polyester and B-481 Chemical Resistant Polyester
Specifications
Certifications/Compliance RoHS 2005/618/EC, MCV RoHS 2002/95/EC
Adhesive Type Non-adhesive
Color Black
Format Cartridge (Ribbon)
Material Polyester
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Equipment) BBP30;BBP31;BBP33;BBP35;BBP37;i3300;S3000;S3100;i5300;S3700
Min. Operating Temperature (Metric) 5° C
Model B30-R6400
No. per Pack 1/Pk.
Quantity 1 Ea.
Thickness (English) 0.0002 in.
Thickness (Metric) 0.0045 mm
Dimensions (L x W) 60 m x 110 mm
Length (English) 200 ft.
Length (Metric) 60 m
Width (English) 4.33 in.
Width (Metric) 110 mm
Alternate Part Number B30-R6400