Learn More
Brady™ 6000 Series Thermal Transfer Printer Ribbon IP™ Printer Enabled
Product Code. 11503035
Change view
Click to view available options
Diameter (English):
1 in.
Length (English):
984 ft.
Width (English):
2.36 in.
3.27 in.
4.33 in.
Unit Size:
300m
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Length (English) Width (English) unitSize
11503035 1 in. 984 ft. 2.36 in. 300m
12849868 1 in. 984 ft. 3.27 in. Each
11895261 1 in. 984 ft. 4.33 in. 300m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11503035 Supplier Brady™ Supplier No. 066202
  • ONLINE EXCLUSIVE PRICE
    133.00 EUR
    130.16 EUR / 300m
    Save 2.84 EUR
    2% Off
View all promotions
In Stock
Add to basket
Add to basket
Includes
  • Spindle
6000 Series Thermal Transfer Printer Ribbon IP™ Printer Enabled is high density ribbon with excellent solvent resistance, high heat resistance, and super scratch resistance.
  • One-color ribbon for printing a single color onto a label
  • R6000 Series Halogen-Free Resin Formulation offers excellent solvent, scratch and heat resistance when printed on the appropriate label material
  • Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra Aggressive Polyester and B-423 Permanent Polyester from Brady™
  • Combine Brady label materials with their recommended ribbon series for superior legend performance
Compliance
RoHS compatibility: Pass 2002/95/EC
Specifications
Certifications/Compliance RoHS 2002/95/EC
Adhesive Type Non-adhesive
Color Black
Finish Gloss
Format Label
Material Resin
Printable Sides Single-side Printable
Print Technology None
Product Type Thermal Transfer Printer Label
Sterility Non-sterile
For Use With (Equipment) IP300 and IP600 printers
Includes Spindle
Quantity 1 Ea.
Size (English) 984 ft. x 2.36 in.
Dimensions (L x W) 299.92 m x 59.94 mm (984 ft. x 2.36 in.)
Length (English) 984 ft.
Length (Metric) 299.92 m
Width (English) 2.36 in.
Width (Metric) 6 cm
Diameter (English) 1 in.
Diameter (Metric) 2.54 cm
Alternate Part Number IP-R6000HF