Learn More
Tocris Bioscience™ BRACO 19 trihydrochloride
Description
BRACO 19 trihydrochloride is a telomerase inhibitor (IC50 = 115 nM). Inhibits expression of human telomerase reverse transcriptase (hTERT), induces cellular senescence and inhibits growth of uterine cancer cells in vitro. Inhibits growth of uterine tumor xenografts in mice.
Specifications
Specifications
| Chemical Name or Material | N,N'-[9[[4-(Dimethylamino)phenyl]amino]-3,6-acridinediyl]bis-1-pyrrolidinepropanamide trihydrochloride |
| Synonym | Telomerase inhibitor |
| Quantity | 50 mg |
| Solubility | Soluble to 50 mM in water and to 50 mM in DMSO |
| Molecular Formula | C35H43N7O2·3HCl |
| Formula Weight | 703.14 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Purity | 94% |
For Research Use Only.