Learn More
Tocris Bioscience™ Bosutinib
GREENER_CHOICE
Product Code. 18015932
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 18015932
Brand: Tocris Bioscience™ 4361/10
Dual Src-Abl inhibitor; antiproliferative
Dual inhibitor of Abl and Src kinases (IC50 = 1.2 nM for Src in an enzymatic assay). Displays antiproliferative activity against chronic myelogenous leukemia (CML) cells and decreases the motility and invasion of breast cancer cell lines. Also exhibits potent antiproliferative activity in anchorage-independent, Src-transformed rat fibroblasts (IC50 = 100 nM). Displays selectivity for Src over non-Src family kinases such as growth factor receptor tyrosine kinases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 380843-75-4
Molecular Formula C26H29Cl2N5O3
Specifications
Quantity 10 mg
Formula Weight 530.45
Purity >99%
Molecular Weight (g/mol) 530.45
Chemical Name or Material Bosutinib