missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Bosutinib

Product Code. 18025112
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18025112

Brand: Tocris Bioscience™ 4361/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dual Src-Abl inhibitor; antiproliferative

Dual inhibitor of Abl and Src kinases (IC50 = 1.2 nM for Src in an enzymatic assay). Displays antiproliferative activity against chronic myelogenous leukemia (CML) cells and decreases the motility and invasion of breast cancer cell lines. Also exhibits potent antiproliferative activity in anchorage-independent, Src-transformed rat fibroblasts (IC50 = 100 nM). Displays selectivity for Src over non-Src family kinases such as growth factor receptor tyrosine kinases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 380843-75-4
Molecular Formula C26H29Cl2N5O3

Specifications

Quantity 50 mg
Formula Weight 530.45
Purity >99%
Molecular Weight (g/mol) 530.45
Chemical Name or Material Bosutinib
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.