Learn More
IKA™ Borosilicate Glass Uncoated Receiving Flask
Product Code. 11855932 Shop All IKA Products
Change view
Click to view available options
Volume (Metric):
250 mL
500 mL
2000 mL
3000 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11855932 500 mL Each
11865932 2000 mL Each
11308121 250 mL Each
11390900 3000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11855932 Supplier IKA™ Supplier No. 0003742400
In Stock
Specifications
Material Borosilicate Glass
Type Receiving Flask
For Use With (Equipment) RV 10.102 Evaporator
Quantity 1 Each
Volume (Metric) 500 mL