Learn More
Fisherbrand™ Borosilicate Glass Tall Form Beakers
Product Code. 15409093 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
150 mL
250 mL
400 mL
600 mL
800 mL
1000 mL
2000 mL
3000 mL
5000 mL
Unit Size:
Each
Pack of 10
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15409093 100 mL Pack of 10
15499083 50 mL Pack of 10
11517412 50 mL Pack of 10
15419093 150 mL Pack of 10
15429093 250 mL Pack of 10
15439093 400 mL Pack of 10
15449093 600 mL Pack of 10
15459093 800 mL Pack of 10
15469093 1000 mL Pack of 10
11597412 1000 mL Each
15479093 2000 mL Each
15489093 3000 mL Each
11912213 5000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15409093
In Stock
Add to basket
Edge
Add to basket
Includes graduations and a spout for easy pouring. Fisherbrand™ Borosilicate Glass Tall Form Beakers are available in a variety of sizes.
  • Borosilicate Glass 3.3
  • Tall Form Graduated with Spout
  • Manufactured to ISO 3819
Specifications
Color Translucent
Diameter (Metric) Outer 48 mm
Capacity (Metric) 100 mL
Material Borosilicate Glass (3.3)
Finish Matte
Height (Metric) 80 mm
Includes Spout
Quantity 10 Pk.
Type Beaker
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Certifications/Compliance ISO 3819
Scale White Graduated Scale
Style Tall Form