Learn More
Pyrex™ Borosilicate Glass Spare Glass Keys
Product Code. 12474761
Change view
Click to view available options
Plug Bore:
4 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Bore Unit Size
12474761 4 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12474761 Supplier Pyrex Supplier No. SSD104K
  • ONLINE EXCLUSIVE PRICE
    30.65 EUR
    25.34 EUR / Pack of 1
    Save 5.31 EUR
    17% Off
Estimated Shipment: 01-02-2027
Manufactured from borosilicate glass
  • Replacement keys for SSD: series stopcocks
  • Manufactured from borosilicate glass
  • Supplied complete with key retaining device
Specifications
Type Spare Glass Key
Plug Bore 4 mm
Diameter (Metric) 14.5 mm
Material Borosilicate Glass