Learn More
Scientific Glass Labs™ Borosilicate Glass Powder Funnel
Product Code. 12903601 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric)Top:
50mm
70mm
100mm
Diameter (Metric) Stem:
13mm
16mm
22mm
Length (Metric) Stem:
20mm
25mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top Diameter (Metric) Stem Length (Metric) Stem Unit Size
12903601 100mm 22mm 25mm Each
12993591 70mm 16mm 25mm Each
12983591 50mm 13mm 20mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12903601
Estimated Shipment: 04-01-2027
Manufactured from borosilicate glass. Scientific Glass Labs™ Borosilicate Glass Powder Funnel is less likely to clog or cake when funneling powdery substances.
  • Manufactured from borosilicate glass.
  • Short stem funnel
  • Less likely to clog or cake when funneling powdery substances
Specifications
Diameter (Metric)Top 100mm
Diameter (Metric) Stem 22mm
Material Borosilicate Glass
Length (Metric) Stem 25mm