Learn More
Fisherbrand™ Glass Square Coverslips
Product Code. 12323128 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Thickness (Metric):
#1 (0.13 to 0.16 mm)
#1.5 (0.16 to 0.19 mm)
Length (Metric):
18 mm
20 mm
22 mm
24 mm
Width (Metric):
18 mm
20 mm
22 mm
24 mm
Unit Size:
Pack of 200
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thickness (Metric) Length (Metric) Width (Metric) Unit Size
12323128 #1 (0.13 to 0.16 mm) 18 mm 18 mm Pack of 200
12343138 #1.5 (0.16 to 0.19 mm) 18 mm 18 mm Pack of 200
12353138 #1.5 (0.16 to 0.19 mm) 20 mm 20 mm Pack of 200
12333128 #1 (0.13 to 0.16 mm) 22 mm 22 mm Pack of 200
12363138 #1.5 (0.16 to 0.19 mm) 22 mm 22 mm Pack of 200
11338503 #1 (0.13 to 0.16 mm) 24 mm 24 mm Pack of 200
12393138 #1.5 (0.16 to 0.19 mm) 24 mm 24 mm Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 12323128
  • ONLINE EXCLUSIVE PRICE
    10.66 EUR
    2.69 EUR / Pack of 200
    Save 7.97 EUR
    74% Off
In Stock
Add to basket
Edge
Add to basket
Glass Square Coverslips are available in a variety of sizes. Packed in plastic boxes to prevent breakage.
  • Manufactured from clear soda glass
  • Consistent in cut, thickness and dimensional accuracy
  • Free from blisters and flaws
Specifications
Product Type Coverslip
Shape Square
Material Soda Lime Glass
Length (Metric) 18 mm
Width (Metric) 18 mm
Thickness (Metric) #1 (0.13 to 0.16 mm)
Quantity 200/Pk.