Learn More
Fisherbrand™ Test Tubes, Borosilicate Glass 3.3, rimless, heavy wall
Product Code. 15892275 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric):
16 mm
Length (Metric):
125 mm
150 mm
Capacity (Metric):
14 mL
17 mL
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) Capacity (Metric) Unit Size
15892275 16 mm 125 mm 14 mL Pack of 100
15862295 16 mm 150 mm 17 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15892275
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
Manufactured from borosilicate glass with a heavy wall design for strong resistance to heat and chemicals. Fisherbrand™ Borosilicate Glass Heavy Wall Rimless Test Tubes are ideal for chemical and biological work.
  • Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for this work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
  • Manufactured from borosilicate glass 3.3
  • Thick-walled and therefore mechanically very resistant
  • With round bottom
Specifications
Certifications/Compliance ISO 4142
Capacity (Metric) 14 mL
Length (Metric) 125 mm
Material Borosilicate Glass
Diameter (Metric) 16 mm
Wall Thickness 1.8 mm