Learn More
Lenz Laborglasinstrumente™ Gas Washing Bottles without Head
Product Code. 10548681
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10548681 100 mL Each
10444491 250 mL Each
10130391 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10548681 Supplier Lenz Laborglasinstrumente™ Supplier No. 05515037
  • ONLINE EXCLUSIVE PRICE
    28.00 EUR
    19.20 EUR / Pack of 1
    Save 8.80 EUR
    31% Off
View all promotions
Estimated Shipment: 07-01-2027
Provides an inexpensive but effective method for the washing and drying of gases. Lenz Laborglasinstrumente™ Gas Washing Bottles without Head are manufactured from durable DURAN™ borosilicate glass.
  • Acc. to DIN 12596, without head
  • With socket NS 29/32
  • Available in a range of capacities
  • Corrosion-free and resistant to thermal shock
Specifications
Capacity (Metric) 100 mL
Material Borosilicate Glass
Type Gas Washing Bottle without Head
Includes Socket
Standard Taper Joint NS 29/32
Height (Metric) 200 mm
Certifications/Compliance DIN 12463
Quantity 1 Pack