Learn More
Fisherbrand™ Borosilicate Glass Class AS Burette with Teflon Tap
Product Code. 10450893 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10450893 25 mL Each
10738081 50 mL Each
11566913 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10450893
Estimated Shipment: 03-05-2027
Add to basket
Add to basket
Complies with BS-IS 385 standards. Fisherbrand™ Borosilicate Glass Class AS Burette with Teflon Tap includes single bore with interchangeable PTFE stopcock. Blue ceramic graduations make volume assessment easy.
  • Constructed of borosilicate glass
  • Single bore with interchangeable PTFE stopcock
  • Blue ceramic graduations
  • Class AS, BS-ISO 385 Compliance
Specifications
Capacity (Metric) 25 mL
Material Borosilicate Glass
Graduations 0.1 mL
Includes Single Bore, Interchangeable Stopcock
Scale Blue
Stopcock Material PTFE
Tolerance ±0.05 mL
Type Buret
Class Class AS
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.