Learn More
Kimble Chase™ Borosilicate Glass Chromatography Column with PTFE Stopcock Assembly
Product Code. 11857413
Change view
Click to view available options
Length (Metric):
500 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
11857413 500 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11857413 Supplier Kimble Chase™ Supplier No. 1780019500
 Restricted Item
Estimated Shipment: 10-11-2026
Specifications
USP Type Type I
Column Capacity 135 mL
Length (Metric) 500 mm
Material Borosilicate Glass
Includes With PTFE stopcock assembly
Temperature (Metric) Range 0 to 100°C
Quantity 1 Each
For Use With (Application) Ion exchange chromatography
Type Chromatography Column
Diameter (Metric) Inner 19 mm