Learn More
Pyrex™ Borosilicate Glass Automatic Burette with Intermediate Stopcock
Product Code. 12842104
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Graduations:
0.02 mL
0.05 mL
0.1 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Graduations Unit Size
12842104 50 mL 0.1 mL Each
12832104 25 mL 0.05 mL Each
12822104 10 mL 0.02 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12842104 Supplier Pyrex Supplier No. 3496/06AS
  • ONLINE EXCLUSIVE PRICE
    266.70 EUR
    230.69 EUR / Pack of 1
    Save 36.01 EUR
    13% Off
Estimated Shipment: 23-02-2027
  • As 3396 range with intermediate stopcock
  • Intermediate stopcock facilitates easy drainage of liquid back into reservoir
  • 1.5mm bore PTFE key stopcock
Specifications
Capacity (Metric) 50 mL
Graduations 0.1 mL