Learn More
Pyrex™ Borosilicate Glass Automatic Burette with Intermediate Stopcock
Product Code. 12832104
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Graduations:
0.02 mL
0.05 mL
0.1 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Graduations unitSize
12832104 25 mL 0.05 mL Each
12842104 50 mL 0.1 mL Each
12822104 10 mL 0.02 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12832104 Supplier Pyrex Supplier No. 3496/04AS
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
  • As 3396 range with intermediate stopcock
  • Intermediate stopcock facilitates easy drainage of liquid back into reservoir
  • 1.5mm bore PTFE key stopcock
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 25 mL
Graduations 0.05 mL