Learn More
Witeg™ Borosilicate glass 3.3 Round bottomed flasks
Product Code. 11837870
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11837870 25 mL Pack of 10
11847870 10 mL Pack of 10
10286373 1000 mL Each
10144283 500 mL Each
10498803 250 mL Each
10743844 100 mL Each
10246233 2000 mL -
10184183 50 mL Each
10792494 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 11837870 Supplier Witeg™ Supplier No. 0 600 025
  • ONLINE EXCLUSIVE PRICE
    87.36 EUR
    80.18 EUR / Pack of 10
    Save 7.18 EUR
    8% Off
Estimated Shipment: 18-09-2026
Specifications
Certifications/Compliance DIN 12348
Capacity (Metric) 25 mL
Material Borosilicate Glass (3.3)
Bottom Shape Round