missing translation for 'onlineSavingsMsg'
Learn More

Boron, Sigma-Aldrich™

Product Code. 15807890
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15807890 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15807890 Supplier Sigma Aldrich Supplier No. 1120700025
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Powder

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. Boron compounds are used as Lewis acid catalysts in organic synthesis, enabling efficient and selective chemical transformations, offering a greener, potentially more sustainable option for advanced synthetic chemistry.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-42-8
Melting Point 2075°C
Boiling Point 4000°C/1013 hPa
Density 1.73 g/cm3 at 15°C
Quantity 25 g
Form Powder
MDL Number MFCD00134034
Synonym Boron, Boron
Molecular Weight (g/mol) 10.81
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.