missing translation for 'onlineSavingsMsg'
Learn More

Boric Acid, TRC

Product Code. 18062320
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
250g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18062320 500 g 500g
18000440 50 g 50g
18000830 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18062320 Supplier Toronto Research Chemicals Supplier No. TRCB675405500G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula B H3 O3
Molecular Weight (g/mol) 61.83
Synonym Boric acid, Boric acid (H3BO3) (6CI, 8CI, 9CI, ACI), Basilit B, Boracic acid, BorActin, Boric acid, Boric acid (B(OH)3), Borofax, Boron hydroxide (B(OH)3), Boron trihydroxide, Bushwhacker, CB BORiD, Dia Flea-Mate, Dr.'s 1 Flea Terminator DF, Dr.'s 1 Flea Terminator DFPBO, Dr.'s 1 Flea Terminator DT, Dr.'s 1 Flea Terminator DTPBO, Entimaden, Flea Prufe, Niban, Niban Granular Bait, Niban-FG, Nitrate balancer, NSC 81726, Optibor, Optibor TP, Orthoboric acid, Orthoboric acid (B(OH)3), Orthoboric acid (H3BO3), PS 1002, Roach Away, Roach Prufe, Super Flea Eliminator, Trihydroxyborane
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Quantity 500 g
InChI Formula InChI=1S/BH3O3/c2-1(3)4/h2-4H
Formula Weight 62.0175
Chemical Name or Material Boric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.