Learn More
Bohlender™ BOLA™ Connectors for Sampling Bags
Product Code. 17126934
Change view
Click to view available options
Material:
PTFE
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
17126934 PTFE Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17126934 Supplier Bohlender™ Supplier No. A22606
  • ONLINE EXCLUSIVE PRICE
    26.84 EUR
    19.60 EUR / Pack of 2
    Save 7.24 EUR
    26% Off
Estimated Shipment: 16-12-2026
PTFE connectors for sampling bags.
  • Connectors to be screwed in the ports of sampling bags
  • Bore diameter: 3 mm
Specifications
Material PTFE
Diameter (English) 0.24 in.
Diameter (Metric) 6 mm