Learn More
Velp™ BOD Sensor Check
Product Code. 11781864 Shop All Velp Products
Change view
Click to view available options
For Use With (Equipment):
BOD/BOD EVO
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
11781864 BOD/BOD EVO Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11781864 Supplier Velp™ Supplier No. A00000135
  • ONLINE EXCLUSIVE PRICE
    149.00 EUR
    147.50 EUR / Pack of 1
    Save 1.50 EUR
    1% Off
Estimated Shipment: 30-11-2026
Velp™ BOD Sensor Check for use with BOD and BOD EVO Sensors.
  • For use with BOD Sensor and BOD EVO Sensor
Specifications
Sensor Types Dissolved Oxygen
For Use With (Equipment) BOD/BOD EVO
Type Dissolved Oxygen Sensor
For Use With (Application) Testing the accuracy of BOD instruments
Dissolved Oxygen (Concentration Range) 0 to 90 mg O2/L for 400 mL; 0 to 250 mg O2/L for 250 mL; 0 to 600 mg O2/L for 150 mL; 0 to 999 mg O2/L for 100 mL; 0 to 4000 mg O2/L for 25 mL
Temperature (Metric) Operating 40°C