Learn More
Boc-Arg(Z)2-OH, TRC
Specifications
Specifications
| Chemical Name or Material | Boc-Arg(Z)2-OH |
| CAS | 51219-19-3 |
| Molecular Formula | C27H34N4O8 |
| Quantity | 500 mg |
| Synonym | Ornithine, N2,N5-dicarboxy-N5-(carboxyamidino)-, dibenzyl tert-butyl ester (7CI), 11-Oxa-2,4,9-triazatridecanoic acid, 8-carboxy-3-imino-12,12-dimethyl-10-oxo-4-[(phenylmethoxy)carbonyl]-, 1-(phenylmethyl) ester, (S)- |
| InChI Formula | InChI=1S/C27H34N4O8/c1-27(2,3)39-25(35)29-21(22(32)33)15-10-16-31(26(36)38-18-20-13-8-5-9-14-20)23(28)30-24(34)37-17-19-11-6-4-7-12-19/h4-9,11-14,21H,10,15-18H2,1-3H3,(H,29,35)(H,32,33)(H2,28,30,34)/t21-/m0/s1 |
| SMILES | CC(C)(C)OC(=O)N[C@@H](CCCN(C(=N)NC(=O)OCc1ccccc1)C(=O)OCc2ccccc2)C(=O)O |
| IUPAC Name | (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-[phenylmethoxycarbonyl-(N-phenylmethoxycarbonylcarbamimidoyl)amino]pentanoic acid |
| Molecular Weight (g/mol) | 542.58 |
| Formula Weight | 542.2377 |