missing translation for 'onlineSavingsMsg'
Learn More

BMX Antibody [PE], Novus Biologicals™

Product Code. 30754217 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30754217 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30754217 Supplier Novus Biologicals Supplier No. AF5887P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

BMX Polyclonal antibody specifically detects BMX in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMX
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias BMX non-receptor tyrosine kinase, Bone marrow tyrosine kinase gene in chromosome X protein, EC 2.7.10, EC 2.7.10.2, Epithelial and endothelial tyrosine kinase, Etk/Bmx cytosolic tyrosine kinase, ETKcytoplasmic tyrosine-protein kinase BMX, NTK38, NTK38 tyrosine kinase, PSCTK2, PSCTK3
Host Species Sheep
Immunogen E. coli-derived recombinant human BMX, Met1-Gln215, Accession # P51813
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 660
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.