Learn More
Tocris Bioscience™ BMS 566419
Description
BMS 566419 is a potent inosine monophosphate dehydrogenase (IMPDH) inhibitor (IC50 = 17 nM). Inhibits proliferation of human T-lymphoblasts and PBMCs in vitro. Reduces paw swelling in a rat adjuvant arthritis model. Orally available.
Specifications
Specifications
| Chemical Name or Material | N-[1-[6-(4-Ethyl-1-piperazinyl)-3-pyridinyl]-1-methylethyl]-2-fluoro-9,10-dihydro-9-oxo-3-acridinecarboxamide |
| CAS | 566161-24-8 |
| Quantity | 50 mg |
| Target | Dehydrogenase Inhibitors |
| Molecular Formula | C28H30FN5O2 |
| Formula Weight | 487.57 |
| Purity | >98% |