Learn More
BMS-303141, MedChemExpress
Product Code. 16493037
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493037 50 mg 50mg
16479527 5 mg 5mg
16426988 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16493037 Supplier Medchem Express Supplier No. HY16107
MedChemExpress BMS-303141 is a potent, cell-permeable ATP-citrate lyase (ACL) inhibitor with an IC50 of 0.13 μM.
BMS-303141 is a potent, cell-permeable ATP-citrate lyase (ACL) inhibitor with an IC50 of 0.13 μM.
Specifications
Chemical Name or Material BMS-303141
CAS 943962-47-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H15Cl2NO4S
Quantity 50 mg
Solubility Information DMSO : 25 mg/mL (58.92 mM; Need ultrasonic)
SMILES O=S(C1=CC(Cl)=CC(Cl)=C1O)(NC2=CC(C3=CC=CC=C3)=CC=C2OC)=O
Molecular Weight (g/mol) 424.3
Formula Weight 424.3
Percent Purity 98.47%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism