Learn More
Berkshire™ BlueSorb™ 750
Product Code. 17897124 Shop All Berkshire Products
Change view
Click to view available options
Dimensions (L x W):
30 x 30 cm
Unit Size:
Pack of 3000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
17897124 30 x 30 cm Pack of 3000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17897124 Supplier Berkshire™ Supplier No. BS750.1212.20
  • ONLINE EXCLUSIVE PRICE
    893.55 EUR
    664.03 EUR / Pack of 3000
    Save 229.52 EUR
    25% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
BlueSorb™ 750 is absorbent with low extractable levels and particle counts. Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose. Blue color allows for differentiation in critical areas.
  • Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
  • Blue color allows differentiation for guarding against migration into critical areas
  • Smooth, absorbent and durable
  • Low extractable levels and particle counts
Specifications
Product Type Surface Wiper
Length (English) 12 in.
Length (Metric) 30 cm
Width (Metric) 30 cm
Material Polyester Cellulose
Color Blue
Dimensions (L x W) 30 x 30 cm
For Use With (Application) ISO Class 5-8