Learn More
De Buyer™ Blowtorches
Product Code. 11982129
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11982129 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11982129 Supplier De Buyer™ Supplier No. 2040.00N
  • ONLINE EXCLUSIVE PRICE
    65.98 EUR
    43.00 EUR / Pack of 1
    Save 22.98 EUR
    34% Off
Estimated Shipment: 16-12-2026
Order Info
dimProductType
Specifications
Product Type Blow Torch
Height (Metric) 157 mm
Includes Automatic ignition button, butane gas cylinder
Quantity Each