Learn More
Thermo Scientific™ Blocker™ BSA
Product Code. 17992361 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
125 mL
200 mL
3 x 200 mL
Chemical Name or Material:
PBS
TBS
Unit Size:
125mL
200mL
Pack of 3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Chemical Name or Material Unit Size
17992361 3 x 200 mL PBS Pack of 3
11889310 125 mL TBS 125mL
10341944 200 mL PBS 200mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17992361 Supplier Thermo Scientific™ Supplier No. 37525X3
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Blocker BSA is a purified 10% solution of bovine serum albumin for blocking in western blotting, ELISA, IHC, and nucleic acid detection methods.
Blocker BSA is a 10% (w/v) solution of high-quality BSA that is useful for saturating excess protein-binding sites on membranes and microplates in immunoassays. It is most frequently diluted 10-fold (to 1% BSA) in 1X PBS or 1X TBS for initial testing. Blocker BSA is usually more effective than nonfat milk for biotin-avidin systems because it contains a single purified protein that is devoid of endogenous biotin.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material PBS
Description Blocker BSA in PBS (10X)
Concentration 10X
For Use With (Application) Western Blot
Physical Form Liquid
Product Line Blocker
Quantity 3 x 200 mL
Formulation 10% BSA