Learn More
Thermo Scientific™ Blocker™ BSA
Shop All Thermo Scientific Products
Click to view available options
Quantity:
125 mL
200 mL
3 x 200 mL
Chemical Name or Material:
PBS
TBS
Unit Size:
125mL
200mL
Pack of 3
Description
Blocker BSA is a purified 10% solution of bovine serum albumin for blocking in western blotting, ELISA, IHC, and nucleic acid detection methods.
Blocker BSA is a 10% (w/v) solution of high-quality BSA that is useful for saturating excess protein-binding sites on membranes and microplates in immunoassays. It is most frequently diluted 10-fold (to 1% BSA) in 1X PBS or 1X TBS for initial testing. Blocker BSA is usually more effective than nonfat milk for biotin-avidin systems because it contains a single purified protein that is devoid of endogenous biotin.
Specifications
Specifications
| Chemical Name or Material | TBS |
| Description | Blocker BSA in TBS (10X) |
| Concentration | 10% BSA |
| For Use With (Application) | Western Blot |
| Physical Form | Liquid |
| Product Line | Blocker |
| Quantity | 125 mL |