Learn More
BRAND™ BLAUBRAND™ One-mark Glass Bulb Pipets, Class A, USP, Certified
Product Code. 15128264 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
0.5 mL
1 mL
2 mL
3 mL
4 mL
5 mL
7 mL
15 mL
20 mL
25 mL
40 mL
100 mL
Unit Size:
Each
Pack of 12
Pack of 6
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15128264 15 mL Pack of 6
15338207 0.5 mL Pack of 12
15188254 1 mL Pack of 12
15121607 2 mL Pack of 12
15214069 3 mL Pack of 6
15181627 4 mL Pack of 6
15181307 4 mL Each
15197655 5 mL Each
15208918 7 mL Pack of 6
15635118 20 mL Each
15198254 25 mL Pack of 6
15379212 40 mL Pack of 6
15111677 100 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15128264 Supplier BRAND™ Supplier No. 30613
Only null left
Add to basket
Add to basket
High contrast blue enamel represents optimum combination of resistance and readability
  • Made of AR-GLAS™
  • Optimized flow geometry in tip ensures reliable liquid dispensing from pipet
  • Fire-polished edges for more durability
  • Available in all ISO sizes from 0.5 to 100mL
  • Supplied with batch number and an accompanying USP batch certificate in original packaging
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP
Class A
Material AR-GLAS
Sterility Non-sterile
Graduated No
Color Green
Volume (Metric) 15 mL
Length (Metric) 510 mm
Type Bulb Pipet
Disposable Reusable
For Use With (Equipment) Pipet Controller