Learn More
BRAND™ BLAUBRAND™ One-mark Glass Bulb Pipets, Class A, USP, Certified
Product Code. 15121607 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
0.5 mL
1 mL
2 mL
3 mL
4 mL
5 mL
7 mL
15 mL
20 mL
25 mL
40 mL
100 mL
Unit Size:
Each
Pack of 12
Pack of 6
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15121607 2 mL Pack of 12
15338207 0.5 mL Pack of 12
15188254 1 mL Pack of 12
15214069 3 mL Pack of 6
15181627 4 mL Pack of 6
15181307 4 mL Each
15197655 5 mL Each
15208918 7 mL Pack of 6
15128264 15 mL Pack of 6
15635118 20 mL Each
15198254 25 mL Pack of 6
15379212 40 mL Pack of 6
15111677 100 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15121607 Supplier BRAND™ Supplier No. 30603
  • ONLINE EXCLUSIVE PRICE
    95.00 EUR
    69.16 EUR / Pack of 12
    Save 25.84 EUR
    27% Off
Estimated Shipment: 02-02-2027
Add to basket
Add to basket
High contrast blue enamel represents optimum combination of resistance and readability
  • Made of AR-GLAS™
  • Optimized flow geometry in tip ensures reliable liquid dispensing from pipet
  • Fire-polished edges for more durability
  • Available in all ISO sizes from 0.5 to 100mL
  • Supplied with batch number and an accompanying USP batch certificate in original packaging
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP
Class A
Material AR-GLAS
Sterility Non-sterile
Graduated No
Color Orange
Volume (Metric) 2 mL
Length (Metric) 330 mm
Type Bulb Pipet
Disposable Reusable
For Use With (Equipment) Pipet Controller