Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Compact Burettes, with Certificate
Product Code. 15884958 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15884958 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15884958 Supplier BRAND™ Supplier No. 913916
Estimated Shipment: 26-11-2026
Fast to dismantle and easy to clean. BRAND™ Blaubrand™ Borosilicate Glass Compact Burettes are also simple to repair because all individual components replaceable. Delivered with individual conformity certificate.
  • Includes removable PTFE stopcock
  • Fast to dismantle and easy to clean.
  • Simple repair - all individual components replaceable.
  • Borosilicate glass construction (Boro 3.3 to DIN EN ISO 385.)
  • Calibrated to deliver (TD, Ex).
  • Schellbach stripe.
  • Delivered with individual conformity certificate.
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Length (Metric) 800 mm
Material Borosilicate Glass
Tolerance ±0.03 mL
Certifications/Compliance DIN EN ISO 385