Learn More
Bollé Safety Over-the-Glass Safety Glasses
Product Code. 13128766 Shop All Bollé Safety Products
Change view
Click to view available options
Weight (Metric):
33 g
67 g
68 g
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) Unit Size
13128766 33 g Each
11745677 68 g Each
11755677 67 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 13128766 Supplier Bollé Safety Supplier No. OVLITLPSI
  • ONLINE EXCLUSIVE PRICE
    11.13 EUR
    5.68 EUR / Pack of 1
    Save 5.45 EUR
    48% Off
Estimated Shipment: 23-11-2026
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Enjoy guaranteed maximum safety for all, including wearers of prescription glasses, with Bollé Safety™ Over-the-Glass Safety Glasses. Made from polycarbonate in a highly wrap-around and attractive style.
  • Highly wrap-around frame
  • Side, upper and lower protection
TRUSTED_SUSTAINABILITY
Specifications
Product Type Over-the-Glass Safety Glasses
Weight (Metric) 33 g
Lens Material Polycarbonate
Frame Material Polycarbonate