Learn More
Swann-Morton™ BladeFLASK Remover, Non-Sterile
Descrizione
- Safely removes and contains scalpel blades.
- This world-first single-handed scalpel blade remover prevents the dangerous injuries commonly caused by removing blades with fingers, forceps or re-sheathing.
- Ensures optimal safety based on speed and convenience for the end-user, ergonomic simplicity and passive, automatic and single-handed safety principles.
- Enables use of preferred metal scalpel handles.
- Wall-mounted or bench-mounted.
- Improves both patient care and staff safety.
- Puncture-proof sharps container which when it is full, can be disposed of safely without any further exposure to sharps contact.
- Auto shut-off to prevent over-filling.
- Built-in counter with a shut-off mechanism that activates automatically after 100 blades have been removed.
- No danger of overfilling.
- Improves patient safety, by enabling surgeons to use ergonomic precision steel scalpels.
- Improves staff safety, by ensuring the used scalpel blade is removed with a safe single-handed technique and then safely contained until final disposal.
Specifica
Specifica
| Quantity | 1 Ea. |
| For Use With (Application) | Scalpel blade removal |
| Material | ABS |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?