Learn More
Brady™ Black and Yellow Warning Tape
Product Code. 10112274 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
33 m x 38 mm
33 m x 50 mm
33 m x 75 mm
Unit Size:
1 roll
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
10112274 33 m x 50 mm 1 roll
11761036 33 m x 38 mm 1 roll
11781036 33 m x 75 mm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10112274 Supplier Brady™ Supplier No. 255312
  • ONLINE EXCLUSIVE PRICE
    27.25 EUR
    25.42 EUR / 1 rolls
    Save 1.83 EUR
    6% Off
In Stock
The marking of transport routes must be both flexible and durable, as they are constantly adapted to the changing demands and needs within your facility or warehouse. This Black and Yellow Warning Tape is a self-adhesive warning film for marking pillars, pylons, the edges of walls, and gate arches.
  • The marking of transport routes must be both flexible and durable, as they are constantly adapted to the changing demands and needs within your facility or warehouse.
  • Self-adhesive warning film for marking pillars, pylons, the edges of walls, and gate arches.
Specifications
Alternate Part Number 255312
Description Warning Tape - Yellow / Black
Dimensions (L x W) 33 m x 50 mm
For Use With (Application) Marking pillars, pylons, the edges of walls, gate arch, transport routes
Length (English) 108.3 ft.
Length (Metric) 33 m
Packaging Quantity 1 Roll
Thickness 0.138 mm
Width (English) 1.97 in.
Width (Metric) 50 mm
Adhesive Type Self-adhesive
Color Black, Yellow