Learn More
Fisherbrand™ Black Polypropylene Screw Caps for Glass Specimen Vials
Product Code. 15324799 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Closure Size:
10 mm
15 mm
18 mm
22 mm
28 mm
Unit Size:
Pack of 300
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Unit Size
15324799 28 mm Pack of 300
15304799 18 mm Pack of 300
15314799 22 mm Pack of 300
15384789 10 mm Pack of 300
15394789 15 mm Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 15324799
  • ONLINE EXCLUSIVE PRICE
    47.20 EUR
    28.06 EUR / Pack of 300
    Save 19.14 EUR
    40% Off
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Screw securely into place to seal squat or tall form glass screw neck specimen vials. Fisherbrand™ Black Polypropylene Screw Caps for Glass Specimen Vials are foil lined and available in sizes to fit a variety of tall or squat form vials.
  • Specifically designed for use with Fisherbrand™ Glass Screw Neck Specimen Vials
  • Polypropylene construction with foil lining
  • Fit both tall and squat form vials
  • Available in a variety of sizes to match vial specifications
  • Screw neck specimen vials are suitable for media, diagnostic, storage, display, and sample collection applications
Specifications
Color Black
Material Polypropylene
Closure Type Screw Cap
Closure Size 28 mm