Learn More
Bisphenol A Monosulfate Sodium Salt, TRC
Product Code. 30005746
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005746 10 mg 10mg
30010121 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30005746 Supplier Toronto Research Chemicals Supplier No. TRCB51956010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 847696-37-1
Molecular Formula C15H15NaO5S
Molecular Weight (g/mol) 330.33
Synonym Phenol, 4-[1-(4-hydroxyphenyl)-1-methylethyl]-, 1-(hydrogen sulfate), sodium salt (1:1) (ACI), Phenol, 4-[1-(4-hydroxyphenyl)-1-methylethyl]-, 1-(hydrogen sulfate), monosodium salt (9CI), Sodium 4-[1-(4-hydroxyphenyl)-1-methylethyl]phenyl sulfate (1:1) (ACI), Bisphenol A monosulfate sodium salt
IUPAC Name sodium;[4-[2-(4-hydroxyphenyl)propan-2-yl]phenyl] sulfate
SMILES OC(C=C1)=CC=C1C(C)(C)C2=CC=C(OS(=O)(O[Na])=O)C=C2
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C15H16O5S.Na/c1-15(2,11-3-7-13(16)8-4-11)12-5-9-14(10-6-12)20-21(17,18)19;/h3-10,16H,1-2H3,(H,17,18,19);/q;+1/p-1
Formula Weight 330.0538
Percent Purity >95%
Chemical Name or Material Bisphenol A Monosulfate Sodium Salt
Grade HPLC