Learn More
Bisphenol A Bissulfate Disodium Salt, TRC
Product Code. 30133990
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133990 250 mg 250mg
30012445 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30133990 Supplier Toronto Research Chemicals Supplier No. TRCB519515250MG
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10040-44-5
Molecular Formula C15 H14 O8 S2 . 2 Na
Molecular Weight (g/mol) 432.38
Synonym Phenol, 4,4'-(1-methylethylidene)bis-, 1,1'-bis(hydrogen sulfate), sodium salt (1:2), Phenol, 4,4'-(1-methylethylidene)bis-, bis(hydrogen sulfate), disodium salt (9CI), Phenol, 4,4'-isopropylidenedi-, bis(hydrogen sulfate) disodium salt (8CI), Sodium S,S'-[(1-methylethylidene)di-4,1-phenylene] disulfate (2:1), Bisphenol A Bissulfate Disodium Salt
IUPAC Name disodium;[4-[2-(4-sulfonatooxyphenyl)propan-2-yl]phenyl] sulfate
SMILES [Na+].[Na+].CC(C)(c1ccc(OS(=O)(=O)[O-])cc1)c2ccc(OS(=O)(=O)[O-])cc2
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C15H16O8S2.2Na/c1-15(2,11-3-7-13(8-4-11)22-24(16,17)18)12-5-9-14(10-6-12)23-25(19,20)21;;/h3-10H,1-2H3,(H,16,17,18)(H,19,20,21);;/q;2*+1/p-2
Formula Weight 431.9925
Chemical Name or Material Bisphenol A Bissulfate Disodium Salt