Learn More
Bismuth Indium Tin ingot (Field's metal)
Product Code. 15412797
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15412797 100 g 100g
15422797 500 g 500g
15432797 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15412797 Supplier Thermo Scientific Alfa Aesar Supplier No. 046895.22
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Bismuth Indium Tin ingot (Field's metal) is used for die casting and rapid prototyping. It acts as an alternative to Wood's metal which is an eutectic alloy of 50% bismuth, 26.7% lead, 13.3% tin, and 10% cadmium by weight.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bismuth Indium Tin ingot (Field's metal)
Avg. Mol. Wt. or Mol. Wt. Range Bi:In:Sn; 32.5:51:16.5 wt%
Quantity 100 g
Form Ingot
Solubility Information Insoluble in water.