missing translation for 'onlineSavingsMsg'
Learn More

Bismuth Citrate, MP Biomedicals

Product Code. 11472076 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472076 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11472076 Supplier MP Biomedicals Supplier No. 0521230780

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 813-93-4
Molecular Formula C6H5BiO7
Quantity 100 g
Synonym bismuth citrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, bismuth 3+ salt 1:1, bismuth citrate usp:usp, bismuth iii citrate, bismuth citrate usp, bismuth 3+ ion citrate, bismuth, citrato 3-, bismuth iii citrate 250g, bismuth 2-hydroxypropane-1,2,3-tricarboxylate
InChI Key ANERHPOLUMFRDC-UHFFFAOYSA-K
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.[Bi+3]
IUPAC Name bismuth;2-hydroxypropane-1,2,3-tricarboxylate
Molecular Weight (g/mol) 398.079
PubChem CID 13135
Formula Weight 398.08
Chemical Name or Material Bismuth Citrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.