missing translation for 'onlineSavingsMsg'
Learn More

Bis(trichloromethyl) Carbonate, Sigma-Aldrich™

Product Code. 15849870
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15849870 100 g 100g
15829870 5 g 5g
15839870 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15849870 Supplier Sigma Aldrich Supplier No. 8142830100
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32315-10-9
Grade Research
Molecular Formula (Cl3CO)2CO
Molecular Weight (g/mol) 296.75

Specifications

Melting Point 77°C to 81°C
Density 1.6 g/cm3 at 20°C
Boiling Point 203°C to 206°C/1013 mbar
MDL Number MFCD00062848
Quantity 100 g
Synonym Bis(trichloromethyl) carbonate, Triphosgene, Carbonic acid bistrichlor methyl ester, Bis(trichloromethyl) carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.