missing translation for 'onlineSavingsMsg'
Learn More
Learn More
bis-Benzimide H-33342 Trihydrochloride, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| CAS | 23491-52-3 |
| Molecular Formula | C27H28N6O |
| Quantity | 25 mg |
| Synonym | hoechst 33342, bisbenzimide, hoechst33342, unii-99kzs6cnzx, 99kzs6cnzx, 2'-4-ethoxyphenyl-5-4-methylpiperazin-1-yl-1h,1'h-2,5'-bibenzo d imidazole, 2'-4-ethoxyphenyl-5-4-methyl-1-piperazinyl-2,5'-bi-benzimidazole, 2'-4-ethoxyphenyl-5-4-methyl-1-piperazinyl-2,5'-bi-1h-benzimidazole, 2'-4'-ethoxyphenyl-5-4-methylpiperazin-1-yl-2,5'-bis-1h-benzimidazole trihydrochloride trihydrate, 2'-p-ethoxyphenyl-5-4-methyl-1-piperazinyl-2,5'-bibenzimidazole |
| InChI Key | PRDFBSVERLRRMY-UHFFFAOYSA-N |
| SMILES | CCOC1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)C4=NC5=C(N4)C=C(C=C5)N6CCN(CC6)C |
| IUPAC Name | 2-(4-ethoxyphenyl)-6-[6-(4-methylpiperazin-1-yl)-1H-benzimidazol-2-yl]-1H-benzimidazole |
| Molecular Weight (g/mol) | 452.562 |
| PubChem CID | 1464 |
| ChEBI | CHEBI:51232 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction